CAS RN 78281-59-1
CAS RN 78281-59-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,4'-Dibromobenzophenone, TRC
CAS: 78281-59-1 Molecular Formula: C13 H8 Br2 O Molecular Weight (g/mol): 340.01 Synonym: (2-Bromophenyl)(4-bromophenyl)methanone,2,4'-Dibromobenzophenone IUPAC Name: (2-bromophenyl)-(4-bromophenyl)methanone SMILES: Brc1ccc(cc1)C(=O)c2ccccc2Br