missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 11466964
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize':
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11466964 1 g 1g
11476964 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11466964 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' B21453.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151411-98-2
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.01
MDL Number MFCD00061210
InChI Key SPPLWANVCVTGEQ-UHFFFAOYSA-N
Synonym 2,4,6-trifluorobenzyl bromide, 2-bromomethyl-1,3,5-trifluorobenzene, 2,4,6-trifluorobenzylbromide, benzene, 2-bromomethyl-1,3,5-trifluoro, alpha-bromo-2,4,6-trifluorotoluene, benzene,2-bromomethyl-1,3,5-trifluoro, pubchem4668, acmc-20ahv2, intermediates-zcf02154, 2,4,6-trifluoro benzyl bromide
PubChem CID 2777059
IUPAC Name 2-(bromomethyl)-1,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(CBr)C(F)=C1

Specifications

Refractive Index 1.5015
Quantity 1 g
UN Number UN3265
Beilstein 8763072
Formula Weight 225.01
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluorobenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.