accessibility menu, dialog, popup

UserName

CAS RN 151411-98-2

IUPAC Name: 2-(bromomethyl)-1,3,5-trifluorobenzene
Synonyms: 2-(bromomethyl)-1,3,5-trifluorobenzene
Molecular Weight (g/mol): 225.01
Molecular Formula: C7H4BrF3
InChi Key: SPPLWANVCVTGEQ-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(CBr)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results