Learn More
2,4,6-Trifluorobenzonitrile, 99%, Thermo Scientific™
Product Code. 11318316
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318316 1 g 1g
11328316 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11318316 Supplier Thermo Scientific Alfa Aesar Supplier No. L15525.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 96606-37-0
Molecular Formula C7H2F3N
Molecular Weight (g/mol) 157.095
MDL Number MFCD00042399
InChI Key HTKFGTCCOJIUIK-UHFFFAOYSA-N
Synonym benzonitrile, 2,4,6-trifluoro, 2,4,6-trifluorobenzenecarbonitrile, ncr bf df ff, pubchem1565, acmc-209s7e, ksc488g9f, 2,4,6-trifluoro-benzonitrile, 2,4,6-trifluorobenzonitrile, benzonitrile, 2,4,6-trifluoro-, radical ion 1-9ci
PubChem CID 737177
IUPAC Name 2,4,6-trifluorobenzonitrile
SMILES C1=C(C=C(C(=C1F)C#N)F)F
Specifications
Melting Point 55°C to 58°C
Boiling Point 179°C
Flash Point 92°C (197°F)
Quantity 1 g
UN Number UN3439
Beilstein 5512504
Formula Weight 157.1
Percent Purity 99%
Chemical Name or Material 2,4,6-Trifluorobenzonitrile