accessibility menu, dialog, popup

UserName

FFNF

CAS RN 96606-37-0

IUPAC Name: 2,4,6-trifluorobenzonitrile
Synonyms: 2,4,6-trifluorobenzonitrile
Molecular Weight (g/mol): 157.1
Molecular Formula: C7H2F3N
InChi Key: HTKFGTCCOJIUIK-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C#N)C(F)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results