Learn More
2,4,6-Trifluorobenzonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Trifluorobenzonitrile |
| CAS | 96606-37-0 |
| Molecular Formula | C7 H2 F3 N |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C7H2F3N/c8-4-1-6(9)5(3-11)7(10)2-4/h1-2H |
| SMILES | Fc1cc(F)c(C#N)c(F)c1 |
| IUPAC Name | 2,4,6-trifluorobenzonitrile |
| Molecular Weight (g/mol) | 157.09 |
| Formula Weight | 157.0139 |