Learn More
2,4,6-Tribromo-3-methylaniline, 98+%, Thermo Scientific™
Product Code. 11424144
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424144 100 g 100g
11404144 5 g 5g
11414144 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11424144 Supplier Thermo Scientific Alfa Aesar Supplier No. A12501.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71642-16-5
Molecular Formula C7H6Br3N
Molecular Weight (g/mol) 343.844
MDL Number MFCD00014768
InChI Key KDZKZKWJNMBNAP-UHFFFAOYSA-N
Synonym 3-methyl-2,4,6-tribromoaniline, 2,4,6-tribromo-m-toluidine, 3-methyl-2,4,6-tribrom-anilin, 2,4,6-tribromo-3-methylphenylamine, benzenamine,2,4,6-tribromo-3-methyl, 2,4,6-tris bromanyl-3-methyl-aniline, 2,4,6-tribromo-3-methylaniline, 98+%
PubChem CID 144499
IUPAC Name 2,4,6-tribromo-3-methylaniline
SMILES CC1=C(C(=C(C=C1Br)Br)N)Br
Specifications
Melting Point 99°C to 102°C
Quantity 100 g
Beilstein 3252610
Formula Weight 343.84
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Tribromo-3-methylaniline