accessibility menu, dialog, popup

UserName

CH3BrBrH2NBr

CAS RN 71642-16-5

IUPAC Name: 2,4,6-tribromo-3-methylaniline
Synonyms: 2,4,6-tribromo-3-methylaniline
Molecular Weight (g/mol): 343.84
Molecular Formula: C7H6Br3N
InChi Key: KDZKZKWJNMBNAP-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(Br)C(N)=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results