missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentamethylbenzophenone, 98%, Thermo Scientific™

Product Code. 11452188
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452188 25 g 25g
11442188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452188 Supplier Thermo Scientific Alfa Aesar Supplier No. A15182.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20386-33-8
Molecular Formula C18H20O
Molecular Weight (g/mol) 252.36
MDL Number MFCD00051595
InChI Key JFFVBXWGARSRSC-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentamethylbenzophenone, 2,3,4,5,6-pentamethylphenyl phenyl methanone, pentamethylphenyl phenyl methanone, 2,3,4,5,6-pentamethyl benzopheneone, acmc-20aoiy, maybridge1_002590, methanone, pentamethylphenyl phenyl-9ci, 2,3,4,5,6-pentamethylphenyl phenyl ketone, 2,3,4,5,6-pentamethylphenyl-phenylmethanone, 2,3,4,5,6-pentamethylphenyl phenyl methanone #
PubChem CID 519768
IUPAC Name (2,3,4,5,6-pentamethylphenyl)-phenylmethanone
SMILES CC1=C(C)C(C)=C(C(=O)C2=CC=CC=C2)C(C)=C1C

Specifications

Melting Point 137°C to 138°C
Quantity 25 g
Formula Weight 252.36
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentamethylbenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.