accessibility menu, dialog, popup

UserName

CH3CH3CH3H3CH3CO

CAS RN 20386-33-8

IUPAC Name: (2,3,4,5,6-pentamethylphenyl)(phenyl)methanone
Synonyms: (2,3,4,5,6-pentamethylphenyl)(phenyl)methanone
Molecular Weight (g/mol): 252.36
Molecular Formula: C18H20O
InChi Key: JFFVBXWGARSRSC-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C(C(=O)C2=CC=CC=C2)C(C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results