missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyethyl)phenol, TRC

Product Code. 30481833
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481833 250 mg 250mg
30469060 100 mg 100mg
30469964 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481833 Supplier Toronto Research Chemicals Supplier No. TRCM262665250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330976-39-1
Molecular Formula C9 H12 O2
Molecular Weight (g/mol) 152.19
Synonym Phenol, 2-(2-methoxyethyl)-, 2-(2-Methoxyethyl)phenol
IUPAC Name 2-(2-methoxyethyl)phenol
SMILES COCCc1ccccc1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H12O2/c1-11-7-6-8-4-2-3-5-9(8)10/h2-5,10H,6-7H2,1H3
Formula Weight 152.0837
Chemical Name or Material 2-(2-Methoxyethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.