CAS RN 330976-39-1
CAS RN 330976-39-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-(2-Methoxyethyl)phenol, TRC
CAS: 330976-39-1 Molecular Formula: C9 H12 O2 Molecular Weight (g/mol): 152.19 Synonym: Phenol, 2-(2-methoxyethyl)-,2-(2-Methoxyethyl)phenol IUPAC Name: 2-(2-methoxyethyl)phenol SMILES: COCCc1ccccc1O
2
2-Methoxyethylphenol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.