missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-4-isopropyl-1,3-benzodioxole, TRC

Product Code. 30384790
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384790 25 mg
30371756 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384790 Supplier Toronto Research Chemicals Supplier No. TRCD47420025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201166-22-5
Molecular Formula C12 H16 O2
Molecular Weight (g/mol) 192.25
Synonym 2,2-Dimethyl-4-(1-methylethyl)-1,3-benzodioxole, Propofol Imp. L (EP)
IUPAC Name 2,2-dimethyl-4-propan-2-yl-1,3-benzodioxole
SMILES CC(C)c1cccc2OC(C)(C)Oc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H16O2/c1-8(2)9-6-5-7-10-11(9)14-12(3,4)13-10/h5-8H,1-4H3
Formula Weight 192.115
Chemical Name or Material 2,2-Dimethyl-4-(1-methylethyl)-1,3-benzodioxole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.