CAS RN 201166-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2,2-Dimethyl-4-isopropyl-1,3-benzodioxole, TRC
CAS: 201166-22-5 Molecular Formula: C12 H16 O2 Molecular Weight (g/mol): 192.25 Synonym: 2,2-Dimethyl-4-(1-methylethyl)-1,3-benzodioxole,Propofol Imp. L (EP) IUPAC Name: 2,2-dimethyl-4-propan-2-yl-1,3-benzodioxole SMILES: CC(C)c1cccc2OC(C)(C)Oc12
2
2,2-Dimethyl-4-(1-methylethyl)-1,3-benzodioxole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.