Learn More
2,2-Dichloroethanol, TRC
Product Code. 30371437
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371437 5 g 5g
30382561 1 g -
30374595 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30371437 Supplier Toronto Research Chemicals Supplier No. TRCD4343305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 598-38-9
Molecular Formula C2 H4 Cl2 O
Molecular Weight (g/mol) 114.96
Synonym 2,2-Dichloroethanol, Dichloroethanol, NSC 60513
IUPAC Name 2,2-dichloroethanol
SMILES OCC(Cl)Cl
Specifications
Quantity 5 g
InChI Formula InChI=1S/C2H4Cl2O/c3-2(4)1-5/h2,5H,1H2
Formula Weight 113.9639
Chemical Name or Material 2,2-Dichloroethanol