CAS RN 598-38-9
CAS RN 598-38-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2-Dichloroethanol, TRC
CAS: 598-38-9 Molecular Formula: C2 H4 Cl2 O Molecular Weight (g/mol): 114.96 Synonym: 2,2-Dichloroethanol,Dichloroethanol,NSC 60513 IUPAC Name: 2,2-dichloroethanol SMILES: OCC(Cl)Cl