Learn More
2,2,5-Trimethyl-3-hexen-1-ol, 97%, Thermo Scientific™
Product Code. 15449099
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449099 1 g 1g
15459099 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15449099 Supplier Thermo Scientific Alfa Aesar Supplier No. L19702.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 608534-29-8
Molecular Formula C9H18O
MDL Number MFCD03788753
Specifications
Refractive Index 1.4435
Quantity 1 g
Beilstein 6774184
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 2,2,5-Trimethyl-3-hexen-1-ol