CAS RN 608534-29-8
CAS RN 608534-29-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2,5-Trimethyl-3-hexen-1-ol, 97%, Thermo Scientific™
CAS: 608534-29-8 Molecular Formula: C9H18O MDL Number: MFCD03788753