Product Code. 17912765
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
17912765
250 mL
250mL
17984685
1 L
1 L
17900765
50 mL
50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
17912765
Supplier
Toronto Research Chemicals
Supplier No.
TRCT796535250ML
Please sign in to purchase this item. Need a web account? Register with us today!
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Chemical Identifiers
CAS
540-84-1
Molecular Formula
C8 H18
Molecular Weight (g/mol)
114.23
Synonym
Pentane, 2,2,4-trimethyl-, 2,2,4-Trimethylpentane, 2,4,4-Trimethylpentane, Isobutyltrimethylmethane, Isooctane, NSC 39117, iso-Octane
IUPAC Name
2,2,4-trimethylpentane
SMILES
CC(C)CC(C)(C)C
Specifications
Quantity
250 mL
InChI Formula
InChI=1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3
Formula Weight
114.1409
Chemical Name or Material
2,2,4-Trimethylpentane
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?