missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ≥99%, Reagent Grade, Solstice

Product Code. 15692790 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
20 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15692790 20 L Glass Bottle 20 L
15602800 500 mL Glass Bottle 500mL
15672790 1 L Glass Bottle 1 L
15682790 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15692790 Supplier Honeywell Research Chemicals Supplier No. 25877620L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point -107°C
Color Colorless
Density 0.692g/mL at 25°C
Quantity 20 L
Explosion Limit 0.06
Flash Point −12°C (10.4°F)
Ignition Point 745°F
Packaging Glass Bottle
Physical Form Liquid
Linear Formula (CH3)2CHCH2C(CH3)3
Vapor Pressure 41mmHg (21°C)
UN Number UN1262
Beilstein 1696876
Boiling Point 98°C to 99°C
Formula Weight 114.23g/mol
Percent Purity ≥99%
Grade ReagentPlus
Chemical Name or Material 2,2,4-Trimethylpentane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.