missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ≥99.5%, Solstice

Product Code. 15626110
Change view
Click to view available options
Quantity:
1 L
20 L
Packaging:
Glass Bottle
Unit Size:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15626110 20 L Glass Bottle 20 L
15616110 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15626110 Supplier Honeywell Riedel-de Haën Supplier No. 65043920L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Density 0.692g/mL at 25°C
Quantity 20 L
Explosion Limit 6%
Ignition Point 745°C
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2C(CH3)3
Vapor Pressure 41mmHg (21°C)
UN Number UN1262
Beilstein 1696876
Formula Weight 114.23g/mol
Percent Purity ≥99.5%
Chemical Name or Material 2,2,4-Trimethylpentane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.