missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,4-triazole-3-sulfonamide, TRC

Product Code. 30135415
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135415 50 mg 50mg
30134855 100 mg 100mg
30006987 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135415 Supplier Toronto Research Chemicals Supplier No. TRCH95511850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89517-96-4
Molecular Formula C2 H4 N4 O2 S
Molecular Weight (g/mol) 148.14
Synonym 1H-1,2,4-Triazole-3-sulfonamide (9CI), s-Triazole-3-sulfonamide (6CI)
IUPAC Name 4H-1,2,4-triazole-3-sulfonamide
SMILES NS(=O)(=O)c1nnc[nH]1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C2H4N4O2S/c3-9(7,8)2-4-1-5-6-2/h1H,(H2,3,7,8)(H,4,5,6)
Formula Weight 148.0055
Chemical Name or Material s-Triazole-3-sulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.