CAS RN 89517-96-4
CAS RN 89517-96-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1H-1,2,4-triazole-3-sulfonamide, TRC
CAS: 89517-96-4 Molecular Formula: C2 H4 N4 O2 S Molecular Weight (g/mol): 148.14 Synonym: 1H-1,2,4-Triazole-3-sulfonamide (9CI),s-Triazole-3-sulfonamide (6CI) IUPAC Name: 4H-1,2,4-triazole-3-sulfonamide SMILES: NS(=O)(=O)c1nnc[nH]1