missing translation for 'onlineSavingsMsg'
Learn More

15-Lipoxygenase 2 Antibody (027), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18215029 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215029 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215029 Supplier Novus Biologicals Supplier No. NBP290287AF750
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

15-Lipoxygenase 2 Monoclonal specifically detects 15-Lipoxygenase 2 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 15-Lipoxygenase 2
Applications ELISA
Classification Monoclonal
Clone 27
Conjugate Alexa Fluor 750
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias 15-lipoxygenase 2, 15-LOX-B, arachidonate 15-lipoxygenase 2, arachidonate 15-lipoxygenase B, Arachidonate 15-lipoxygenase type II, arachidonate 15-lipoxygenase, second type, arachidonate 15-lipoxygenase, type B, arachidonate omega(6) lipoxygenase, EC 1.13.11, EC 1.13.11.33,15-LOX-215S-lipoxygenase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human 15-Lipoxygenase 2 (AAH35217.1; Met1-Ile676).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 247
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.