missing translation for 'onlineSavingsMsg'
Learn More

Chlorzoxazone-4,6,7-d3, TRC

Product Code. 17987939
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17987939 10 mg 10mg
17998209 1 mg 1mg
17909229 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17987939 Supplier Toronto Research Chemicals Supplier No. TRCC42870210MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185173-60-7
Molecular Formula C7 D3 H Cl N O2
Molecular Weight (g/mol) 172.584
Synonym 5-Chloro-4,6,7-trideuterio-1,3-benzoxazol-2-ol, 5-Chloro-1,3-benzoxazol-2(3H)-one-4,6,7-d3, 5-Chloro-2-benzoxazolinone-4,6,7-d3, Chlorzoxazone-d3, Chlorzoxazone-4,6,7-D3
IUPAC Name 5-chloro-4,6,7-trideuterio-3H-1,3-benzoxazol-2-one
SMILES [2H]c1c([2H])c2OC(=O)Nc2c([2H])c1Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H4ClNO2/c8-4-1-2-6-5(3-4)9-7(10)11-6/h1-3H,(H,9,10)/i1D,2D,3D
Formula Weight 172.012
Chemical Name or Material Chlorzoxazone-4,6,7-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.