CAS RN 1185173-60-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Chlorzoxazone-4,6,7-d3, TRC
CAS: 1185173-60-7 Molecular Formula: C7 D3 H Cl N O2 Molecular Weight (g/mol): 172.584 Synonym: 5-Chloro-4,6,7-trideuterio-1,3-benzoxazol-2-ol,5-Chloro-1,3-benzoxazol-2(3H)-one-4,6,7-d3,5-Chloro-2-benzoxazolinone-4,6,7-d3,Chlorzoxazone-d3,Chlorzoxazone-4,6,7-D3 IUPAC Name: 5-chloro-4,6,7-trideuterio-3H-1,3-benzoxazol-2-one SMILES: [2H]c1c([2H])c2OC(=O)Nc2c([2H])c1Cl
2
Chlorzoxazone-4,6,7-d3, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.