missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsiloxy)cyclohexene, TRC

Product Code. 30455238
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455238 25 g 25g
30453091 10 g 10g
30455000 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455238 Supplier Toronto Research Chemicals Supplier No. TRCT79694525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6651-36-1
Molecular Formula C9 H18 O Si
Molecular Weight (g/mol) 170.324
Synonym 1-[(Trimethylsilyl)oxy]cyclohexene, (1-Cyclohexen-1-yloxy)trimethylsilane, (Cyclohex-1-enyloxy)trimethylsilane, 1-(Trimethylsiloxy)-1-cyclohexene, 1-(Trimethylsilyloxy)-1-cyclohexene, 1-(Trimethylsilyloxy)cyclohexene, 1-Cyclohexenol Trimethylsilyl Ether, 1-Cyclohexenyl Trimethylsilyl Ether, Cyclohexanone Enol Trimethylsilyl Ether, Cyclohexanone Trimethylsilyl Enol Ether, α-(Trimethylsiloxy)cyclohexene
IUPAC Name cyclohexen-1-yloxy(trimethyl)silane
SMILES C[Si](C)(C)OC1=CCCCC1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H18OSi/c1-11(2,3)10-9-7-5-4-6-8-9/h7H,4-6,8H2,1-3H3
Formula Weight 170.113
Chemical Name or Material 1-(Trimethylsiloxy)cyclohexene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.