accessibility menu, dialog, popup

UserName

H3CSiH3CCH3O

CAS RN 6651-36-1

IUPAC Name: (cyclohex-1-en-1-yloxy)trimethylsilane
Synonyms: (cyclohex-1-en-1-yloxy)trimethylsilane
Molecular Weight (g/mol): 170.33
Molecular Formula: C9H18OSi
InChi Key: SBEMOANGDSSPJY-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC1=CCCCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)