Learn More
1-Naphthyl butyrate, 98+%
Product Code. 11440827
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440827 1 g 1g
11450827 5 g 5g
11460827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11440827 Supplier Thermo Scientific Alfa Aesar Supplier No. B23221.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 3121-70-8 | C14H14O2 | 214.264 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3121-70-8
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00003923
InChI Key XIVJNRXPRQKFRZ-UHFFFAOYSA-N
Synonym 1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik
PubChem CID 76571
IUPAC Name naphthalen-1-yl butanoate
SMILES CCCC(=O)OC1=CC=CC2=CC=CC=C21
Specifications
Density 1.09
Boiling Point 125°C to 130°C (1 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.571
Quantity 1 g
Beilstein 2558646
Formula Weight 214.27
Percent Purity ≥98%
Chemical Name or Material 1-Naphthyl butyrate