accessibility menu, dialog, popup

UserName

CAS RN 3121-70-8

H3COO

CAS RN 3121-70-8

IUPAC Name: naphthalen-1-yl butanoate
Synonyms: naphthalen-1-yl butanoate
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: XIVJNRXPRQKFRZ-UHFFFAOYSA-N
SMILES: CCCC(=O)OC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)