missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylimidazole, Sigma-Aldrich™
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product has been enhanced for catalysis. Find details here.
Specifications
Specifications
| Melting Point | -2°C |
| Density | 1.035 g/cm3 at 20°C |
| Boiling Point | 198°C/1013 hPa |
| MDL Number | MFCD00005292 |
| Quantity | 1 L |
| Synonym | 1-Methylimidazole, N-Methylimidazole, 1-Methylimidazole |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?