accessibility menu, dialog, popup

UserName

CH3NN

CAS RN 616-47-7

IUPAC Name: 1-methyl-1H-imidazole
Synonyms: 1-methylimidazole
Molecular Weight (g/mol): 82.11
Molecular Formula: C4H6N2
InChi Key: MCTWTZJPVLRJOU-UHFFFAOYSA-N
SMILES: CN1C=CN=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (10)
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)
  • (10)
  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (6)
  • (5)
  • (2)
  • (2)