missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentyl-4-nitrosopiperazine, TRC

Product Code. 18072120
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18072120 100 mg 100mg
18070620 1 g 1g
17909839 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18072120 Supplier Toronto Research Chemicals Supplier No. TRCC213795100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61379-66-6
Molecular Formula C9 H17 N3 O
Molecular Weight (g/mol) 183.25
Synonym Piperazine, 1-cyclopentyl-4-nitroso- (9CI, ACI), 1-Cyclopentyl-4-nitrosopiperazine (ACI)
IUPAC Name 1-cyclopentyl-4-nitroso-piperazine
SMILES O=NN1CCN(CC1)C2CCCC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H17N3O/c13-10-12-7-5-11(6-8-12)9-3-1-2-4-9/h9H,1-8H2
Formula Weight 183.1372
Percent Purity >95%
Chemical Name or Material 1-Cyclopentyl-4-nitrosopiperazine
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.