CAS RN 61379-66-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-Cyclopentyl-4-nitrosopiperazine, TRC
CAS: 61379-66-6 Molecular Formula: C9 H17 N3 O Molecular Weight (g/mol): 183.25 Synonym: Piperazine, 1-cyclopentyl-4-nitroso- (9CI, ACI),1-Cyclopentyl-4-nitrosopiperazine (ACI) IUPAC Name: 1-cyclopentyl-4-nitroso-piperazine SMILES: O=NN1CCN(CC1)C2CCCC2