Learn More
1-Butylpyrrolidine, TRC
Product Code. 30137378
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137378 500 mg 500mg
30134649 100 mg 100mg
30007111 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137378 Supplier Toronto Research Chemicals Supplier No. TRCB700948500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 767-10-2
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
Synonym N-Butylpyrrolidine, N-Butyltetrahydropyrrole
IUPAC Name 1-butylpyrrolidine
SMILES CCCCN1CCCC1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C8H17N/c1-2-3-6-9-7-4-5-8-9/h2-8H2,1H3
Formula Weight 127.1361
Chemical Name or Material 1-Butylpyrrolidine