CAS RN 767-10-2
CAS RN 767-10-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Butylpyrrolidine, TRC
CAS: 767-10-2 Molecular Formula: C8H17N Molecular Weight (g/mol): 127.23 Synonym: N-Butylpyrrolidine,N-Butyltetrahydropyrrole IUPAC Name: 1-butylpyrrolidine SMILES: CCCCN1CCCC1