missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-dichloro-3-fluorobenzene, 97%, Thermo Scientific Chemicals

Product Code. 11309355
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309355 1 g 1g
11319355 5 g 5g
11329355 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309355 Supplier Thermo Scientific Alfa Aesar Supplier No. L16713.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 202865-57-4 | Molecular Formula: C6H2BrCl2F | Molecular Weight: 243.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-57-4
Molecular Formula C6H2BrCl2F
Molecular Weight (g/mol) 243.88
MDL Number MFCD00142585
InChI Key CAYJMDVKWMVOLG-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-fluorobromobenzene, 3-bromo-2,5-dichloro-1-fluorobenzene, pubchem3211, acmc-1cm10, #, 2-bromo-6-fluoro-1,4-phenylene dichloride, benzene, 1-bromo-2,5-dichloro-3-fluoro, 1-bromanyl-2,5-bis chloranyl-3-fluoranyl-benzene
PubChem CID 618513
IUPAC Name 1-bromo-2,5-dichloro-3-fluorobenzene
SMILES FC1=CC(Cl)=CC(Br)=C1Cl

Specifications

Refractive Index 1.5757
Quantity 1 g
Formula Weight 243.89
Percent Purity 97%
Chemical Name or Material 1-Bromo-2,5-dichloro-3-fluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.