accessibility menu, dialog, popup

UserName

BrClFCl

CAS RN 202865-57-4

IUPAC Name: 1-bromo-2,5-dichloro-3-fluorobenzene
Synonyms: 1-bromo-2,5-dichloro-3-fluorobenzene
Molecular Weight (g/mol): 243.88
Molecular Formula: C6H2BrCl2F
InChi Key: CAYJMDVKWMVOLG-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(Br)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results