missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopropane-1-Carboxylic Acid, MP Biomedicals™

Product Code. 11493151 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493151 1 g 1g
11473151 100 mg 100mg
11483151 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493151 Supplier MP Biomedicals Supplier No. 0215034891
  • ONLINE EXCLUSIVE PRICE
    758.00 EUR
    721.35 EUR / 1g
    Save 36.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 22059-21-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00009944
InChI Key PAJPWUMXBYXFCZ-UHFFFAOYSA-N
ChEBI CHEBI:18053
IUPAC Name 1-aminocyclopropane-1-carboxylic acid
SMILES NC1(CC1)C(O)=O

Specifications

Color White
Quantity 1 g
Synonym 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
PubChem CID 535
Formula Weight 101.1
Physical Form Solid
Chemical Name or Material 1-Aminocyclopropane-1-Carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.