accessibility menu, dialog, popup

UserName

CAS RN 22059-21-8

IUPAC Name: 1-aminocyclopropane-1-carboxylic acid
Synonyms: 1-aminocyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: PAJPWUMXBYXFCZ-UHFFFAOYSA-N
SMILES: NC1(CC1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (4)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (7)
  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)