missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanamine Sulfate, TRC

Product Code. 30008766
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008766 2.5 g 2.5g
30006554 500 mg 500mg
30004912 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008766 Supplier Toronto Research Chemicals Supplier No. TRCA5760102.5G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31377-23-8
Molecular Formula 2(C10H17N) . H2SO4
Synonym Amantadine Sulfate, Infex, PK-Merz, Tricyclo[3.3.1.13,7]decan-1-amine Sulfate
SMILES NC12CC3CC(CC(C3)C1)C2.NC45CC6CC(CC(C6)C4)C5.OS(=O)(=O)O

Specifications

Quantity 2.5 g
Formula Weight 400.2396
Chemical Name or Material 1-Adamantanamine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.