accessibility menu, dialog, popup

UserName

CAS RN 31377-23-8

NH2NH2OHSOHOO

CAS RN 31377-23-8

IUPAC Name: bis(adamantan-1-amine); sulfuric acid
Synonyms: bis(amantadine); sulfuric acid
Molecular Weight (g/mol): 400.58
Molecular Formula: C20H36N2O4S
InChi Key: MYWTWSQFJLXGGQ-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC12CC3CC(CC(C3)C1)C2.NC12CC3CC(CC(C3)C1)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results