missing translation for 'onlineSavingsMsg'
Learn More

1-(9H-Carbazol-4-yloxy)-3-(propan-2-ylamino)propan-2-ol, TRC

Product Code. 18003700
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18003700 5 mg 5mg
30040417 25 mg 25mg
30040929 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18003700 Supplier Toronto Research Chemicals Supplier No. TRCB4166035MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57775-29-8
Molecular Formula C18 H22 N2 O2
Molecular Weight (g/mol) 298.38
Synonym 1-(9H-Carbazol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol, (R,S)-Carazolol, (±)-Carazolol, BM 51052, Carazolol, Conducton, DL-Carazolol, Racemic Carazolol, Suacron, Suacron Praemix
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(O)COc1cccc2[nH]c3ccccc3c12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H22N2O2/c1-12(2)19-10-13(21)11-22-17-9-5-8-16-18(17)14-6-3-4-7-15(14)20-16/h3-9,12-13,19-21H,10-11H2,1-2H3
Formula Weight 298.1681
Percent Purity >95%
Chemical Name or Material Carazolol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.