accessibility menu, dialog, popup

UserName

H3CCH3NHOHONH

CAS RN 57775-29-8

IUPAC Name: 1-(9H-carbazol-4-yloxy)-3-[(propan-2-yl)amino]propan-2-ol
Synonyms: (+)-carazolol carazololum carazolol, (+-)-isomer carazolol suacron [veterinary] (-)-carazolol carazolol, (+)-isomer (+-)-carazolol suacron carazolol, (-)-
Molecular Weight (g/mol): 298.39
Molecular Formula: C18H22N2O2
InChi Key: BQXQGZPYHWWCEB-UHFFFAOYNA-N
SMILES: CC(C)NCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)