missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dichloronaphthalene, TRC

Product Code. 30365345
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365345 25 mg 25mg
30354430 10 mg 10mg
30353484 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365345 Supplier Toronto Research Chemicals Supplier No. TRCD26999525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-74-0
Molecular Formula C10 H6 Cl2
Molecular Weight (g/mol) 197.06
Synonym Naphthalene, 1,8-dichloro-, 1,8-Dichloronaphthalene, PCN 9, 1,8-DiCN, 1,8-DCN, PCN-9
IUPAC Name 1,8-dichloronaphthalene
SMILES Clc1cccc2cccc(Cl)c12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H6Cl2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H
Formula Weight 195.9847
Chemical Name or Material 1,8-Dichloronaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.