CAS RN 2050-74-0
CAS RN 2050-74-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,8-Dichloronaphthalene 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
1,8-Dichloronaphthalene, TRC
CAS: 2050-74-0 Molecular Formula: C10 H6 Cl2 Molecular Weight (g/mol): 197.06 Synonym: Naphthalene, 1,8-dichloro-,1,8-Dichloronaphthalene,PCN 9,1,8-DiCN,1,8-DCN,PCN-9 IUPAC Name: 1,8-dichloronaphthalene SMILES: Clc1cccc2cccc(Cl)c12