missing translation for 'onlineSavingsMsg'
Learn More

1,8,9-Trihydroxyanthracene, 97%, Thermo Scientific™

Product Code. p-7025009
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469816 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11469816 Supplier Thermo Scientific Alfa Aesar Supplier No. B20303.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Dithranol was used as a matrix in MALDI TOF mass spectrometry measurements of sol-gel films for DNA microarray applications. It was also used as matrix in matrix-assisted laser desorption/ionization-mass spectrometry (MALDI-MS) and liquid secondary ion-tandem mass spectrometry (LSI-MS/MS) for the analysis of synthetic polymers to generate values for the average mass and the mass of the end groups. It was also used in MALDI-MS for direct detection of condensed tannins in bark. It is successfully used for the treatment of psoriasis.

Solubility
Insoluble in water. Soluble in acetic acid, chloroform (20 mg/mL), acetone, benzene, solutions of alkali hydroxides, fixed oil. Slightly soluble in 95% ethanol, alcohol, ether, and glacial acetic acid.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1143-38-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00053409
InChI Key NUZWLKWWNNJHPT-UHFFFAOYSA-N
Synonym anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy
PubChem CID 2202
ChEBI CHEBI:37510
IUPAC Name 1,8-dihydroxy-9,10-dihydroanthracen-9-one
SMILES OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2

Specifications

Melting Point 177°C to 181°C
Quantity 5 g
Beilstein 2054360
Merck Index 14,684
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 1,8,9-Trihydroxyanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.