accessibility menu, dialog, popup

UserName

OHOHO

CAS RN 1143-38-0

IUPAC Name: 1,8-dihydroxy-9,10-dihydroanthracen-9-one
Synonyms: anthralin chrysodermol anthra-derm dithranolum cigthranol batridol dithranol ccris 628 anthraderm psoriacid-stift
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: NUZWLKWWNNJHPT-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (4)
  • (1)
Physical Form 
  • (3)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)