Learn More
1-(5-Methyl-2-pyridyl)piperazine, 97%, Thermo Scientific™
Product Code. 11499600
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499600 250 mg 250mg
11409610 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11499600 Supplier Thermo Scientific Alfa Aesar Supplier No. H50897.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104395-86-0
Molecular Formula C10H15N3
Molecular Weight (g/mol) 177.25
MDL Number MFCD08061050
InChI Key QDRKPHJFMGUXPN-UHFFFAOYSA-N
Synonym 1-5-methyl-2-pyridinyl piperazine, 1-5-methylpyridin-2-yl piperazine, 1-5-methyl-2-pyridyl piperazine, piperazine, 1-5-methyl-2-pyridinyl, 5-methyl-2-pyridyl piperazine, 1-5-methyl-pyridin-2-yl-piperazine, piperazine,1-5-methyl-2-pyridinyl, 1-5-m ethyl-pyridin-2-yl-piperazine
PubChem CID 11480779
IUPAC Name 1-(5-methylpyridin-2-yl)piperazine
SMILES CC1=CN=C(C=C1)N1CCNCC1
Specifications
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material 1-(5-Methyl-2-pyridyl)piperazine