accessibility menu, dialog, popup

UserName

CH3NNNH

CAS RN 104395-86-0

IUPAC Name: 1-(5-methylpyridin-2-yl)piperazine
Synonyms: 1-(5-methylpyridin-2-yl)piperazine
Molecular Weight (g/mol): 177.25
Molecular Formula: C10H15N3
InChi Key: QDRKPHJFMGUXPN-UHFFFAOYSA-N
SMILES: CC1=CN=C(C=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results